Learn More
Medchemexpress LLC HY-N2292 5mg Medchemexpress, Kinsenoside CAS:151870-74-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN22925MG
Encompass_Preferred
Medchemexpress, HY-N2292 5mg Kinsenoside CAS:151870-74-5 Kinsenoside is a main active component isolated from plants of the genus Anoectochilus, and exhibits many biological activities and pharmacological effects. Kinsenoside rescues the nucleus pulposus cells (NPCs) viability under oxidative stress and protects against apoptosis, senescence and mitochondrial dysfunction in a Nrf2-dependent way. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2884