Learn More
Grainger KIPP Adjustable Handle Ball Knob
Supplier:  Grainger 3DEW7
Encompass_Preferred
The KIPP Adjustable Handle Ball Knob is a black plastic ball knob with a threaded stud that can be attached to machines, fixtures, or other equipment to provide a comfortable grip for hand operation. It has a ball diameter of 1-1/4" and the stud has a thread size of M10 x 1.5. The stud length is 1-1/2" and the overall length including the knob is 2-3/4". This knob provides tool-free adjustment with a dual-component design - the knob rotates on the threaded stud to provide comfortable positioning. It is made from a fiberglass reinforced plastic with a matte black finish. The temperature resistance is up to 266°F.
Catalog No. NC2880027