Learn More
Medchemexpress LLC HY-19708 10mg Medchemexpress, KIRA6 CAS:1589527-65-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1970810MG
Encompass_Preferred
Medchemexpress, HY-19708 10mg KIRA6 CAS:1589527-65-0 KIRA6 is an advanced small-molecule IRE1α RNase kinase inhibitor with an IC50 of 0.6 µM[2]. KIRA6 can trigger an apoptotic response. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9288