Learn More
Cayman Chemical KIspeptIn-10mouserat 10mg
SDP
Supplier:  Cayman Chemical 1403610 MG
Encompass_Preferred
A neuropeptide and kisspeptin receptor ligand; corresponds to amino acids 110-119 of rodent KiSS1; inhibits FGF2-induced migration of NIH3T3 cells expressing the mouse kisspeptin receptor at 10 µM; increases serum FSH and LH levels in adult male rats (ED50s = 400 and 4 pmol/animal, i.c.v.)
Catalog No. 50-295-3798