Learn More
Cayman Chemical KIspeptIn-10zebrafIshtr 10mg
SDP
Supplier:  Cayman Chemical 3565210 MG
Encompass_Preferred
A neuropeptide; increases levels of the mRNA encoding Lh receptor, as well as Pra and Prb, in isolated zebrafish ovarian follicles at 10 ng/ml; increases luteinizing hormone serum levels in female, but not male, goldfish at 0.1 and 1 UG/g
Catalog No. 50-291-0547