Learn More
Medchemexpress LLC Kiss2 peptide acetate | 1111070-93-9 | 1257.44 | 5 MG
Supplier: Medchemexpress LLC HYP10256A5MG
Kiss2 peptide acetate is the acetate form of Kiss2 peptide (HY-P10256). It acts as a positive regulator of reproduction.
- Binds with its cognate receptor Kiss2R (GPR54) in COS-7 cells
- Activates PKA and PKC signaling pathways through Gas and Gaq proteins
- Enhances the activity of cAMP response element-dependent luciferase (CRE-luc) and serum response element-dependent luciferase (SRE-luc)