Learn More
Lonza Walkersville PYROSPERSE Dispersing Agent
Supplier:  Lonza Walkersville N188
Encompass
PYROSPERSE (Dispersing Agent) helps eliminate endotoxin binding or masking in some samples - solving problems of inhibitory behavior. Examples of samples that may show endotoxin binding behavior include plasma protein fractions, electrolyte solutions, and lipid emulsions. PYROSPERSE may be used with any of our LAL Kits.Benefits:Endotoxin and functionality testedContent:5 mL/vial
Catalog No. NC9699852