Learn More
Chemglass Life Sciences Certified Kit, Preassembled, Clear with Spot, 2.0mL 12x32mm 9mm Thread Large Opening Vial and 0.040" PTFE/Silicone Septa Lined Bonded Cap
SDP
Supplier:  Chemglass Life Sciences CV5145CERT
Encompass_Preferred
Certified Kit, Preassembled, Clear with Spot, 2.0mL 12x32mm 9mm Thread Large Opening Vial and 0.040" PTFE/Silicone Septa Lined Bonded Cap
Catalog No. 50-153-2631