Learn More
Perkin Elmer US LLC DSC Sample Calibration Kit
Supplier:  Perkin Elmer US LLC N5190762
Encompass_Preferred
The DSC Sample Calibration Kit includes a small calibration sample of Indium, a foil strip calibration sample of Zinc, an Aluminum Sample Pan Kit (02190041), and a 6 mm Calibration Disc (02510008).
Catalog No. 50-904-4710