Learn More
Molecular Devices Molecular Devices dsDNA Assay Kit, AccuBlue Pico, 5 pg-3 ng
Supplier:  Molecular Devices R8355
Encompass_Preferred
SpectraMax Quant AccuBlue Pico dsDNA Assay Kit is a fluorescence-based kit for sensitive quantitation of double-stranded DNA in 96-well microplate format. The kit uses a high-sensitivity dye and enhancer system optimized for microplate readers and includes reagents and standards needed for multiple plates. Preconfigured protocols are provided for SoftMax Pro software to simplify data acquisition.
  • Detects dsDNA from 5 pg to 3 ng per well.
  • Recommended excitation 468 nm, emission 507 nm.
  • Bulk kit format sufficient for ten 96-well plates.
  • Includes dye, buffer, enhancer, and dsDNA standard.
  • Preconfigured microplate reader protocols for SoftMax Pro software.
Catalog No. 50-000-14359