Learn More
Hach Company PourThru Cell Kit
Supplier:  Hach Company 5940400
Encompass
Permits rapid colorimetric measurements without requiring individual glass sample cells. The Pour-Thru Cell fits into the cell compartment, and solutions being analyzed are poured into the cell by means of a funnel and tubing.
Catalog No. 50-199-7215