Learn More
Molecular Devices Molecular Devices Reporter Assay Kit, Dual Luciferase, 200 Reactions, Explorer
Supplier:  Molecular Devices R8361
Encompass_Preferred
The SpectraMax DuoLuc Reporter Assay Kit is a dual-luciferase system for sensitive quantitation of firefly and Renilla luciferase activity in mammalian cells. The Explorer kit provides reagents for 200 reactions and is optimized for microplate readers with injector systems.
  • Dual-luciferase assay for firefly and Renilla detection.
  • Explorer kit provides reagents for 200 reactions.
  • Flash-type luminescence with sensitivity down to ten cells per well.
  • Includes passive lysis buffer, firefly and Renilla assay buffers, substrates, and coelenterazine.
  • Optimized for microplate readers with injector systems and supported by preconfigured software protocols.
  • Store at -80°C; select components may be stored at -20°C; kit stable for 3 months from receipt.
Catalog No. 50-000-14585