Learn More
Medchemexpress LLC KJ Pyr 9 | 581073-80-5 | 99.6% | 385.37 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY1973510MM/1ML
Encompass_Preferred
KJ Pyr 9 is an inhibitor of MYC with a Kd of 6.5 nM in in vitro assay. It interferes with MYC-MAX complex formation and specifically inhibits MYC-induced oncogenic transformation in cell culture. It preferentially interferes with the proliferation of MYC-overexpressing human and avian cells and reduces the MYC-driven transcriptional signature.
  • Inhibits MYC-MAX complex formation
  • Inhibits MYC-induced oncogenic transformation
  • Preferentially interferes with MYC-overexpressing cell proliferation
  • Reduces MYC-driven transcriptional signature
  • Can halt tumor growth in vivo
Catalog No. 50-004-00892