Learn More
Carbosynth LLC. KKALLALALHHLAHLALHLALALKKA, 1mg, MW: (M.W. 2779.53), Molecular formula: C132H228N38O27
Supplier: Carbosynth LLC. PHR3642PI1MG
Synonyms: H-Lys-Lys-Ala-Leu-Leu-Ala-Leu-Ala-Leu-His-His-Leu-Ala-His-Leu-Ala-Leu-His-Leu-Ala-Leu-Ala-Leu-Lys-Lys-Ala-OH, KKALLALALHHLAHLALHLALALKKA, Application: Amphipathic Peptide Antibiotic, LAH4, Storage Temperature: -20°C, References: A. Kichler, C. Leborgne, J. März, O. Danos, and B. Bechinger, Proc. Natl. Acad. Sci. USA, 100 (4), 1564 (2003).