Learn More
Medchemexpress LLC HY-145597 5mg , KL-11743 CAS:1369452-53-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1455975MG
Encompass_Preferred
HY-145597 5mg KL-11743 CAS: 1369452-53-8 KL-11743 is a potent, orally active, and glucose-competitive inhibitor of the class I glucose transporters, with IC50s of 115, 137, 90, and 68 nM for GLUT1, GLUT2, GLUT3, and GLUT4, respectively. KL-11743 specifically blocks glucose metabolism. KL-11743 can synergize with electron transport inhibitors to induce cell death. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0870