Learn More
Medchemexpress LLC Kl044 | 1801856-93-8 | 99.1% | 50 MG

Supplier: Medchemexpress LLC HY11950650MG
KL044 is a small-molecule stabilizer of the circadian clock protein cryptochrome (CRY) used as a research chemical probe for biochemical and cell-based studies.
- Chemical name: 2-(9H-carbazol-9-yl)-N-(2-chloro-6-cyanophenyl)acetamide.
- Cas number: 1801856-93-8.
- Molecular formula: C21H14ClN3O; molecular weight: 359.81 g/mol.
- Purity: 99.05%.
- Available formulations: solid, solution (10 mM in DMSO), and solid-plus-solvent; multiple milligram sizes available.
- Storage: powder -20°C (up to 3 years) or 4°C (up to 2 years); in solvent -80°C (6 months) or -20°C (1 month).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.