Learn More
ATCC Klebsiella michiganensis Strain 957532 (ATCC BAA-3290)
Supplier:  ATCC BAA3290
Encompass
Klebsiella michiganensis (strain 957532) is a drug-resistant, genome-sequenced clinical isolate supplied for laboratory research applications such as antimicrobial resistance studies, molecular detection assay development, and bioinformatics. This organism is intended for research use only and is not for diagnostic or therapeutic use.
  • Drug-resistant clinical isolate recovered from a wound in South Korea (2013).
  • Genome-sequenced strain suitable for antimicrobial and bioinformatics research.
  • BSL 2 organism intended for laboratory research use only.
  • Growth on trypticase soy agar/broth at 37°C under aerobic conditions.
  • Long-term storage at -80°C or colder.
  • Susceptibility profile: resistant to multiple antibiotics; susceptible to carbapenems and select agents.
Catalog No. NC3538726