Learn More
ATCC Klebsiella pneumoniae; Strain 1100770
Supplier:  ATCC BAA2473
Encompass
Klebsiella pneumoniae strain 1100770 is a whole-genome sequenced bacterium that was isolated from human urine. This strain is an NDM-1 reference strain and is resistant to ertapenem and imipenem. The strain can be used in antimicrobial resistance research, drug development, infectious disease research, urinary tract infection research, and respiratory disease research.
Catalog No. 50-238-3142