Learn More
ATCC Klebsiella pneumoniae; subsp. rhinoscleromatis
Supplier:  ATCC 13884
Encompass
Klebsiella pneumoniae subsp. rhinoscleromatis strain NCTC 5046 [NCDC 417-68, R-70] was isolated in Sumatra from a human nose. This strain is a whole-genome sequenced type strain with applications in infectious disease and respiratory disease research.
Catalog No. 50-238-1802