Learn More
ATCC Klebsiella quasipneumoniae
Supplier:  ATCC 700603
Encompass
Klebsiella quasipneumoniae strain K6 is a whole-genome sequenced bacterium that was isolated from the urine of a hospitalized patient in Richmond, Virginia. This bacterium produces beta-lactamase SHV-18 and can be used as a control for extended-spectrum beta-lactamase production. This organism is a CLSI quality control strain for antimicrobial susceptibility testing.
Catalog No. 50-238-2079
 Restricted Item