Learn More
ATCC Klebsiella variicola; Strain: PS-53
Supplier:  ATCC 31488
Encompass
This bacterial strain was isolated from soil in Panama and is known to produce polysaccharide S-53.
Catalog No. 50-238-3747