Learn More
MSC KleenGuard Lab Coat, Polypropylene, 2X-Large, Non-Hazardous Particulate Protection, 50/Cs.
Supplier:  MSC 07933005
Encompass
This disposable lab coat provides protection against non-hazardous particles in various work environments. Constructed from lightweight, breathable spunbond polypropylene, the garment is designed for comfort during extended wear. It features elastic wrists for a secure fit and a 4-snap closure system.
  • Breathable material allows moisture to escape for enhanced comfort
  • Designed for protection against non-hazardous particles
  • Equipped with elastic cuffs and a 4-snap closure
  • Free from natural rubber latex and silicone
  • Meets ANSI/ISEA 101-1996 standards
  • Recyclable through the RightCycle program
Catalog No. 50-006-85620