Learn More
Immudex Klickmer dextran backbone reagent, 200 μL, 160 nM concentration, mono-biotinylated ligands, ~40 acceptor sites, for research use only
SDP
Supplier:  Immudex DX01K 200
Encompass
Klickmer dextran backbone reagent, 200 μL, 160 nM concentration, mono-biotinylated ligands, ~40 acceptor sites, for research use only
Catalog No. NC2310660