Learn More
Medchemexpress LLC Klotho beta Mouse 10ug | 10ug
SDP
Supplier:  Medchemexpress LLC HYP7871410UG
Encompass_Preferred
Klotho beta protein represses cholesterol 7-alpha-hydroxylase (CYP7A1) a key enzyme in bile acid synthesis It enhances binding between fibroblast growth factor receptors and fibroblast growth factor 21 and directly interacts with FGF19 and FGF21 This positions Klotho beta as a crucial modulator of metabolic and growth factor signaling pathways Klotho beta Protein Mouse (HEK293 His) is the recombinant mouse-derived Klotho beta protein expressed by HEK293 with C-10 His labeled tag
Catalog No. 50-001-94248