Learn More
Medchemexpress LLC Klotho-derived peptide 1 hydrochloride | 98.6% | 3228.44 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP10414A5MG
Encompass_Preferred
Klotho-derived peptide 1 hydrochloride (KP1 human) blocks TGF-β/TGF-β receptor 2 interaction and inhibits TGF-β-induced activation of Smad2/3 and mitogen-activated protein kinase (MAPK). It exhibits anti-fibrotic and kidney protective effects in a mouse model and is intended for research use only.
  • Blocks TGF-β/TGF-β receptor 2 interaction
  • Inhibits TGF-β-induced activation of Smad2/3 and MAPK
  • Exhibits anti-fibrotic effects
  • Provides kidney protective effects
  • Useful in inflammation and immune system disease research
  • Applicable in stem cell/Wnt, TGF-beta/Smad, and MAPK/ERK pathway studies
Catalog No. 50-004-23696