Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC KMG-104 | 852057-94-4 | 99.1% | 435.33 | 1 ML
SDP

Supplier:  Medchemexpress LLC HY13964610MM/1ML

Encompass_Preferred

KMG-104 is a cell-impermeant, highly selective fluorescent Mg2+ probe. It has been widely used to reveal Mg2+ mobilization in the cytoplasm of various cell types. This product is intended for research use only.

  • Cell-impermeant
  • Highly selective fluorescent Mg2+ probe
  • Excited at 490 nm, emitted fluorescence around 510 nm
  • Dissociation constant (Kd) for Mg: 3 mM
  • Soluble at 50 mg/mL in DMSO
  • Appearance: brown to red solid

Catalog No. 50-003-49811


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.