Learn More
Medchemexpress LLC HY-15465 5mg Medchemexpress, KN-93 CAS:139298-40-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY154655MG
Encompass_Preferred
Medchemexpress, HY-15465 5mg KN-93 CAS:139298-40-1 KN-93 is a cell-permeable, reversible and competitive inhibitor calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0632
 Restricted Item