Learn More
Selleck Chemical LLC KN-93 5mg
SDP
Supplier:  Selleck Chemical LLC S67875MG
Encompass_Preferred
KN-93 is a cell-permeable, reversible and competitive inhibitor of Ca2+/calmodulin-dependent protein kinase II (CaMK-II) with Ki of 2.58 ?M at rabbit myocardial CaM kinase activity. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3222