Learn More
Selleck Chemical LLC KN-93 Phosphate 10mg 1188890-41-6
SDP
Supplier:  Selleck Chemical LLC S742310MG
Encompass_Preferred
KN-93 Phosphate is a potent and specific inhibitor of Ca2+/calmodulin-dependent protein kinase II (CaMKII) with Ki of 0.37 ?M, no remarkable inhibitory effects on APK, PKC, MLCK or Ca2+-PDE activities. KN-93 attenuates CaMKII-induced autophagy. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4755