Learn More
Medchemexpress LLC KN-93 (phosphate) | 1913269-12-1 | C26H32ClN2O8PS | 50 MG
Supplier: Medchemexpress LLC HY15465B50MG
KN-93 phosphate is a cell-permeable, reversible, and competitive inhibitor of calmodulin-dependent kinase type II (CaMKII) with a Ki of 370 nM. It is intended for research use only and is not sold to patients. This product typically appears as a white to off-white solid.
- Cell-permeable and reversible
- Specific CaMKII inhibitor
- Induces G1 cell cycle arrest and apoptosis
- Inhibits cell growth stimulated by various growth factors in NIH 3T3 fibroblasts
- Impacts H+, K+-ATPase activity
- Prevents early afterdepolarizations
- High purity (99.72%)