Learn More
TARGETMOL CHEMICALS INC KNK437 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T244425MG
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 200 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. KNK437 (Heat Shock Protein Inhibitor I) a pan-HSP inhibitor suppresses the synthesis of inducible HSPs(HSP105 HSP72 and HSP40). purity: 98%
Catalog No. 50-258-0808