Learn More
Medchemexpress LLC HY-100110 5mg Medchemexpress, KNK437 CAS:218924-25-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1001105MG
Encompass_Preferred
Medchemexpress, HY-100110 5mg KNK437 CAS:218924-25-5 KNK437 is a HSP inhibitor, and inhibits the induction of HSP105, HSP70, and HSP40. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-1716