Learn More
Cayman Chemical KnosamInhydrochlorIde 10mg
SDP
Supplier:  Cayman Chemical 1662310 MG
Encompass_Preferred
An antibiotic that can inhibit cell wall synthesis in plant-pathogenic oomycetes (MIC = 25 UG/ml for P. medicaginis M2913) and certain fungi as well as some bacterial species (MIC = 400 UG/ml for S. aureus)
Catalog No. 50-296-7526