Learn More
Aviva Systems Biology Corp KNTC1 Antibody - N-terminal region (ARP33708_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP33708_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against KNTC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human KNTC1. Alternate names: FLJ36151, KIAA0166, RODApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-55122