Learn More
Medchemexpress LLC Ko 143 | 461054-93-3 | 100.0% | 469.57 | 25MG
SDP
Supplier:  Medchemexpress LLC 25MGHY10010
Encompass_Preferred
Ko 143 is a potent and selective ATP-binding cassette subfamily G member 2 (ABCG2/BCRP) inhibitor. This compound demonstrates over 200-fold selectivity compared to P-gp and MRP-1 transporters.
  • Potent and selective ABCG2/BCRP inhibitor.
  • Shows over 200-fold selectivity compared to P-gp and MRP-1 transporters.
  • Has an EC90 of 26 nM for BCRP.
Catalog No. 50-003-47338