Learn More
ABclonal Technology KO Validated a-Catenin
SDP
Supplier:  ABclonal Technology A19004100UL
Encompass_Preferred
This gene encodes a member of the catenin family of proteins that play an important role in cell adhesion process by connecting cadherins located on the plasma membrane to the actin filaments inside the cell The encoded mechanosensing protein contains three vinculin homology domains and undergoes conformational changes in response to cytoskeletal tension resulting in the reconfiguration of cadherin-actin filament connections Certain mutations in this gene cause butterfly-shaped pigment dystrophy
Catalog No. 50-000-08336