Learn More
ABclonal Technology KO Validated GSK3a Rabbit mAb
SDP
Supplier:  ABclonal Technology A19060100UL
Encompass_Preferred
This gene encodes a multifunctional Ser Thr protein kinase that is implicated in the control of several regulatory proteins including glycogen synthase and transcription factors such as JUN It also plays a role in the WNT and PI3K signaling pathways as well as regulates the production of beta-amyloid peptides associated with Alzheimer s disease
Catalog No. 50-000-08338