Learn More
ABclonal Technology KO Validated HDAC1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2649220UL
Encompass_Preferred
Histone acetylation and deacetylation catalyzed by multisubunit complexes play a key role in the regulation of eukaryotic gene expression The protein encoded by this gene belongs to the histone deacetylase acuc apha family and is a component of the histone deacetylase complex It also interacts with retinoblastoma tumor-suppressor protein and this complex is a key element in the control of cell proliferation and differentiation Together with metastasis-associated protein-2 it deacetylates p53 and modulates its effect on cell growth and apoptosis
Catalog No. 50-000-11414