Learn More
ABclonal Technology KO Validated III-Tubulin
SDP
Supplier:  ABclonal Technology A24067100UL
Encompass_Preferred
This gene encodes a class III member of the beta tubulin protein family Beta tubulins are one of two core protein families (alpha and beta tubulins) that heterodimerize and assemble to form microtubules This protein is primarily expressed in neurons and may be involved in neurogenesis and axon guidance and maintenance Mutations in this gene are the cause of congenital fibrosis of the extraocular muscles type 3 Alternate splicing results in multiple transcript variants A pseudogene of this gene is found on chromosome 6
Catalog No. 50-000-07026