Learn More
ABclonal Technology KO Validated IRF3 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2727220UL
Encompass_Preferred
This gene encodes a member of the interferon regulatory transcription factor (IRF) family The encoded protein is found in an inactive cytoplasmic form that upon serine threonine phosphorylation forms a complex with CREBBP This complex translocates to the nucleus and activates the transcription of interferons alpha and beta as well as other interferon-induced genes The protein plays an important role in the innate immune response against DNA and RNA viruses Mutations in this gene are associated with Encephalopathy acute infection-induced herpes-specific 7
Catalog No. 50-000-09310