Learn More
ABclonal Technology [KO Validated] KRAS Rabbit pAb
SDP
Supplier:  ABclonal Technology A1270420UL
Encompass_Preferred
This gene, a Kirsten ras oncogene homolog from the mammalian ras gene family, encodes a protein that is a member of the small GTPase superfamily. A single amino acid substitution is responsible for an activating mutation. The transforming protein that results is implicated in various malignancies, including lung adenocarcinoma, mucinous adenoma, ductal carcinoma of the pancreas and colorectal carcinoma. Alternative splicing leads to variants encoding two isoforms that differ in the C-terminal region.
Catalog No. 50-001-16699