Learn More
ABclonal Technology KO Validated LKB1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2263620UL
Encompass_Preferred
The protein encoded by this gene is a serine threonine kinase that regulates cell polarity and energy metabolism and functions as a tumor suppressor Mutations in this gene have been associated with the autosomal dominant Peutz-Jeghers syndrome as well as with skin pancreatic and testicular cancers
Catalog No. 50-000-05662