Learn More
ABclonal Technology [KO Validated] NDUFB4 Rabbit pAb
SDP
Supplier:  ABclonal Technology A13820500UL
Encompass_Preferred
This gene encodes a non-catalytic subunit of the multisubunit NADHubiquinone oxidoreductase, the first enzyme complex in the mitochondrial electron transport chain (complex I). Mammalian complex I is composed of 45 different subunits and transfers electrons from NADH to ubiquinone.
Catalog No. 50-000-99681