Learn More
ABclonal Technology KO Validated NQO1 Rabbit mAb
SDP
Supplier:  ABclonal Technology A2229020UL
Encompass_Preferred
This gene is a member of the NAD(P)H dehydrogenase (quinone) family and encodes a cytoplasmic 2-electron reductase This FAD-binding protein forms homodimers and reduces quinones to hydroquinones This protein s enzymatic activity prevents the one electron reduction of quinones that results in the production of radical species Mutations in this gene have been associated with tardive dyskinesia (TD) an increased risk of hematotoxicity after exposure to benzene and susceptibility to various forms of cancer Altered expression of this protein has been seen in many tumors and is also associated with Alzheimer s disease (AD) Alternate transcriptional splice variants encoding different isoforms have been characterized
Catalog No. 50-000-05206