Learn More
ABclonal Technology KO Validated NRF2 Rabbit mAb
SDP
Supplier:  ABclonal Technology A3577100UL
Encompass_Preferred
This gene encodes a transcription factor which is a member of a small family of basic leucine zipper (bZIP) proteins The encoded transcription factor regulates genes which contain antioxidant response elements (ARE) in their promoters many of these genes encode proteins involved in response to injury and inflammation which includes the production of free radicals Multiple transcript variants encoding different isoforms have been characterized for this gene
Catalog No. 50-000-13625