Learn More
ABclonal Technology KO Validated TRIM21/SS-A
SDP
Supplier:  ABclonal Technology A22724100UL
Encompass_Preferred
This gene encodes a member of the tripartite motif (TRIM) family The TRIM motif includes three zinc-binding domains a RING a B-box type 1 and a B-box type 2 and a coiled-coil region The encoded protein is part of the RoSSA ribonucleoprotein which includes a single polypeptide and one of four small RNA molecules The RoSSA particle localizes to both the cytoplasm and the nucleus RoSSA interacts with autoantigens in patients with Sjogren syndrome and systemic lupus erythematosus Alternatively spliced transcript variants for this gene have been described but the full-length nature of only one has been determined
Catalog No. 50-000-05148