Learn More
Medchemexpress LLC HY-15716 10mg Medchemexpress, Kobe0065 CAS:436133-68-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1571610MG
Encompass_Preferred
Medchemexpress, HY-15716 10mg Kobe0065 CAS:436133-68-5 Kobe0065 is a novel and effective inhibitor of Ras-Raf interaction, competitively inhibiting the binding of H-Ras·GTP to c-Raf-1 RBD with a Ki value of 46±13 μM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1776