Learn More
Apexbio Technology LLC Kobe0065 436133-68-5 100mg
SDP
Supplier:  Apexbio Technology LLC B3586100
Encompass_Preferred
Kobe0065 (CAS 436133-68-5) is a small-molecule inhibitor targeting Ras proteins by disrupting the interaction between GTP-bound H-Ras or M-Ras and the Ras-binding domain of c-Raf-1 Identified via in silico screening Kobe0065 inhibits H-Ras GTP c-Raf-1 binding with a Ki of 46 M In NIH 3T3 cells it decreases the association of mutant H-RasG12V with c-Raf-1 in a dose-dependent manner (IC50 10 M) and suppresses downstream phosphorylation of MEK and ERK at 20 M In colony formation assays Kobe0065 inhibits proliferation of H-rasG12V transfected NIH 3T3 cells (IC50 0 5 M) and demonstrates activity against Ras-mutant cancer cell lines In xenograft models administration reduces tumor growth Kobe0065 serves as a research tool for studying Ras-mediated signaling and potential anti-tumor mechanisms
Catalog No. 50-000-91101