Learn More
Medchemexpress LLC Kobe2601 | 316151-68-5 | 99.7% | 5 MG

Supplier: Medchemexpress LLC HY1192305MG
Kobe2601 is a small-molecule research probe that acts as an analogue of Kobe family compounds with reported activity against Ras and Raf. It is intended for in vitro research and is annotated to affect GPCR/G protein and MAPK/ERK signaling pathways.
- Small-molecule Ras and Raf probe for biochemical studies.
- Annotated to modulate GPCR/G protein and MAPK/ERK signaling.
- Available as a 5 mg vial for laboratory use.
- Molecular weight 351.31; formula C13H10FN5O4S.
- Soluble in DMSO for stock solution preparation.
- Intended for research use only; not for human or clinical use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.