Learn More
Electron Microscopy Sciences Kodak Polymax T Developer 1 QT
SDP
Supplier:  Electron Microscopy Sciences 7423212
Encompass_Preferred
Tray processing. Liquid developer-concentrate for use with cold-tone papers. Medium contrast, high capacity, fast acting. Dilute 1:9 for use.
Catalog No. 50-268-06